Learn More
Waters Corp BioSuite Diol (OH) Column, 125Å, 13 µm, 21.5 mm X 300 mm, 5K - 150K, 1/pk
Catalog No. 50780979 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioSuite SEC Columns are available in 4.6 mm, 7.5 mm, and 7.8 mm i.d. analytical and 21.5 mm i.d. preparative and guard columns.
Catalog No. 50-780-979 Supplier Waters Corp Supplier No. 186002169
Add to Cart
Add to Cart
Request a Quote
BioSuite SEC Columns are available in 4.6 mm, 7.5 mm, and 7.8 mm i.d. analytical and 21.5 mm i.d. preparative and guard columns.