Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, Diol (OH), 125Å, 5 μm, 7.8 x 300 mm

Catalog No. 50780974 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This high-performance liquid chromatography (HPLC) column is designed for size exclusion chromatography (SEC). It features diol (OH) chemistry, silica particle substrate, a 125 Å pore size, and a molecular weight separation range of 5,000 to 150,000. It has an inner diameter of 7.8 mm and a length of 300 mm.

  • Diol (OH) chemistry
  • SEC separation mode
  • Silica particle substrate
  • pH range of 2.5 to 7.5
  • Maximum pressure of 1050 psi (72 bar)
  • Endcapped
  • Molecular weight range of 5,000 to 150,000
  • Spherical particle shape
  • Particle size of 5 μm
  • Parker-style endfitting
  • Pore size of 125 Å
  • Column format
  • HPLC system compatibility
  • USP classification L59
  • Inner diameter of 7.8 mm
  • Length of 300 mm
  • Protein application
  • One unit per package

Catalog No. 50-780-974 Supplier Waters Corp Supplier No. 186002164
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This high-performance liquid chromatography (HPLC) column is designed for size exclusion chromatography (SEC). It features diol (OH) chemistry, silica particle substrate, a 125 Å pore size, and a molecular weight separation range of 5,000 to 150,000. It has an inner diameter of 7.8 mm and a length of 300 mm.

  • Diol (OH) chemistry
  • SEC separation mode
  • Silica particle substrate
  • pH range of 2.5 to 7.5
  • Maximum pressure of 1050 psi (72 bar)
  • Endcapped
  • Molecular weight range of 5,000 to 150,000
  • Spherical particle shape
  • Particle size of 5 μm
  • Parker-style endfitting
  • Pore size of 125 Å
  • Column format
  • HPLC system compatibility
  • USP classification L59
  • Inner diameter of 7.8 mm
  • Length of 300 mm
  • Protein application
  • One unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.