Learn More
Waters Corp Waters Diol (OH) Column, 250Å, 10 μm, 7.5 mm × 300 mm
Supplier:  Waters Corp 186002170
Encompass_Preferred
This column is designed for protein and peptide analysis, offering excellent resolution and recovery for biopharmaceutical and diagnostic reagents. It features a 250Å pore size with standard size-exclusion (SEC) packing, utilizing a wettable silica-based material, and is chemically stable from 2.5 to 7.5 pH.
  • Supports legacy methods.
  • Available in multiple internal diameters for analytical and preparative applications.
  • Provides excellent resolution and recovery.
  • Features 250Å pore size with standard size-exclusion (SEC) packing.
  • Chemically stable from 2.5 to 7.5 pH range.
  • Suitable for 10 μm particles in the molecular weight range of 10,000 to 500,000 Daltons.
  • Undergoes extensive testing for quality and reproducibility.
  • Compatible with guard columns for contamination protection.
Catalog No. 50-780-980