Learn More
Waters Corp Waters BioSuite Diol (OH) Column, 250 Å, 13 μm, 21.5 mm X 300 mm
Catalog No. 50780981 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BioSuite Diol (OH) Column supports legacy methods for protein and peptide analysis. It features a 250Å pore size and Standard size-Exclusion (SEC) packing with wettable silica. Chemically stable from 2.5 to 7.5 pH, it offers excellent resolution and recovery, enhancing protein separations and characterization of complex molecules.
  • Designed for protein and peptide analysis.
  • Wettable silica-based material.
  • Chemically stable from 2.5 to 7.5 pH.
  • Excellent resolution and recovery of proteins and peptides.
  • Suitable for isolation, analysis, and characterization.
  • Extensive testing for high quality and reproducibility.
  • Increased separation efficiency.
Catalog No. 50-780-981 Supplier Waters Corp Supplier No. 186002171
Add to Cart
Add to Cart
Request a Quote
This BioSuite Diol (OH) Column supports legacy methods for protein and peptide analysis. It features a 250Å pore size and Standard size-Exclusion (SEC) packing with wettable silica. Chemically stable from 2.5 to 7.5 pH, it offers excellent resolution and recovery, enhancing protein separations and characterization of complex molecules.
  • Designed for protein and peptide analysis.
  • Wettable silica-based material.
  • Chemically stable from 2.5 to 7.5 pH.
  • Excellent resolution and recovery of proteins and peptides.
  • Suitable for isolation, analysis, and characterization.
  • Extensive testing for high quality and reproducibility.
  • Increased separation efficiency.