Learn More
Waters Corp BioSuite Diol (OH) Column, 250Å, 5 µm, 7.8 mm X 300 mm, 10K - 500K, 1/pk
Supplier:  Waters Corp 186002165
Encompass_Preferred
BioSuite SEC Columns are available in 4.6 mm, 7.5 mm, and 7.8 mm i.d. analytical and 21.5 mm i.d. preparative and guard columns.
Catalog No. 50-780-975