Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters BioSuite Diol (OH) Column, 13 μm, 450 Å, 7.5 mm X 300 mm

Catalog No. 50780982 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Diol (OH) Column primarily supports legacy methods and is designed for Size Exclusion Chromatography (SEC) applications. It is available in various inner diameters for analytical and preparative separations.

  • Chemistry: Diol (OH)
  • Separation mode: SEC
  • Particle substrate: Silica
  • pH range: 2.5 to 7.5
  • Maximum pressure: 15 bar (225 psi)
  • Particle size: 13 μm
  • Pore size: 450 Å
  • Inner diameter: 7.5 mm
  • Length: 300 mm
  • USP classification: L59
  • Molecular weight range: 20,000 to 1,000,000
  • System: HPLC
  • Application: Protein

Catalog No. 50-780-982 Supplier Waters Corp Supplier No. 186002172
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Diol (OH) Column primarily supports legacy methods and is designed for Size Exclusion Chromatography (SEC) applications. It is available in various inner diameters for analytical and preparative separations.

  • Chemistry: Diol (OH)
  • Separation mode: SEC
  • Particle substrate: Silica
  • pH range: 2.5 to 7.5
  • Maximum pressure: 15 bar (225 psi)
  • Particle size: 13 μm
  • Pore size: 450 Å
  • Inner diameter: 7.5 mm
  • Length: 300 mm
  • USP classification: L59
  • Molecular weight range: 20,000 to 1,000,000
  • System: HPLC
  • Application: Protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.