Learn More
Waters Corp BioSuite Column, Diol (OH), 450Å, 8 μm
Supplier: Waters Corp 186002166
The BioSuite Diol (OH) Column supports legacy methods and meets demanding requirements for protein and peptide analysis. It provides exceptional resolution and recovery, processing protein separations quickly and aiding in developing next-generation pharmaceuticals.
- Chemistry: Diol (OH)
- Separation mode: SEC (Size-Exclusion Chromatography)
- Particle substrate: Silica
- pH range: 2.5 to 7.5
- Maximum pressure: 525 psi (36 bar)
- Molecular weight range: 20,000 to 7,000,000
- Particle size: 8 μm
- Pore size: 450 Å
- Inner diameter: 7.8 mm
- Length: 300 mm
- Endcapped: Yes
- Endfitting type: Parker-style
- System: HPLC
- USP classification: L59
- Application: Protein analysis and characterization
- Provides exceptional resolution and recovery of proteins and peptides.
- Ensures high quality and reproducibility.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.