Learn More
Waters Corp HPLC Column, C18 PA-B, 3.5 μm, 2.1 × 100 mm
Catalog No. 50781110 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC column is designed for reversed-phase chromatography of proteins and peptides. It features C18 PA-B chemistry, a 3.5 μm particle size, and 2.1 × 100 mm dimensions, suitable for analytical to preparative applications.
  • Features C18 (pC18) chemistry on a pH stable polymeric resin, with 500 Å pore size for proteins up to 2,500,000 Daltons.
  • Provides high chemical stability.
  • Suitable for analytical, preparative, narrow-bore HPLC, and LC/MS applications.
  • Separates proteins, peptides, and other biomolecules via reversed-phase chromatography at elevated pH.
  • Supports characterization and isolation of biotherapeutics; addresses proteomics needs.
  • Supports HPLC and LC/MS analyses of peptides and proteins.
Catalog No. 50-781-110 Supplier Waters Corp Supplier No. 186002434
This HPLC column is designed for reversed-phase chromatography of proteins and peptides. It features C18 PA-B chemistry, a 3.5 μm particle size, and 2.1 × 100 mm dimensions, suitable for analytical to preparative applications.
  • Features C18 (pC18) chemistry on a pH stable polymeric resin, with 500 Å pore size for proteins up to 2,500,000 Daltons.
  • Provides high chemical stability.
  • Suitable for analytical, preparative, narrow-bore HPLC, and LC/MS applications.
  • Separates proteins, peptides, and other biomolecules via reversed-phase chromatography at elevated pH.
  • Supports characterization and isolation of biotherapeutics; addresses proteomics needs.
  • Supports HPLC and LC/MS analyses of peptides and proteins.