Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp BioSuite C18 PA-A Column, 3μm, 2.1 x 100 mm

Supplier:  Waters Corp 186002426

Encompass_Preferred

This product is a BioSuite C18 PA-A column, which is a type of HPLC column.

  • Stationary phase: C18 PA-A
  • Particle size: 3μm
  • Dimensions: 2.1 x 100 mm

Catalog No. 50-781-102


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.