Learn More
Waters Corp Waters Column, C18 PA-A, 3 μm, 4.6 mm x 250 mm, 100 Å
Supplier: Waters Corp 186002432
The BioSuite C18 PA-A Column is a premier reversed-phase column, specifically optimized for peptide mapping in LC or LC-MS applications. It provides excellent batch-to-batch reproducibility and is uniquely QC tested for consistent results.
- C18 chemistry with silica particle substrate.
- Reversed-phase separation mode.
- pH range of 2 to 8.
- Maximum pressure of 6000 psi (415 Bar).
- Spherical particles, 3 μm size, 100 Å pore size.
- 4.6 mm inner diameter, 250 mm length.
- Endcapped for enhanced performance.
- Suitable for molecular weights 1000 to 30000.
- Manufactured under cGMP, ISO 9002 conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.