Learn More
Waters Corp WATERS CORPORATION BioSuite C18 PA-A Column, 3 μm, 100Å, 4.6 mm x 50 mm
Catalog No. 50781105 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioSuite C18 PA-A Column is a silica-based, difunctionally bonded C18 column, featuring an optimal combination of C18 ligand density, silica pore diameter, and endcapping. It is compatible with 100% aqueous mobile phases and stable under low pH conditions, making it ideal for effective separation of complex peptide mixtures.
  • Silica-based, difunctionally bonded C18 chemistry with optimized ligand density.
  • Compatible with 100% aqueous mobile phases and stable under low pH conditions.
  • Designed for effective retention of polar peptides.
  • Performs exceptionally well with LC-MS preferred mobile phases.
  • Offers high peak capacity for peptide mapping and complex sample separation.
  • The 4.6 x 50 mm column can separate 164 peaks with a resolution (Rs) of 1.
  • Suitable for routine and fast separations of 25–35 peptides.
Catalog No. 50-781-105 Supplier Waters Corp Supplier No. 186002429
Add to Cart
Add to Cart
The BioSuite C18 PA-A Column is a silica-based, difunctionally bonded C18 column, featuring an optimal combination of C18 ligand density, silica pore diameter, and endcapping. It is compatible with 100% aqueous mobile phases and stable under low pH conditions, making it ideal for effective separation of complex peptide mixtures.
  • Silica-based, difunctionally bonded C18 chemistry with optimized ligand density.
  • Compatible with 100% aqueous mobile phases and stable under low pH conditions.
  • Designed for effective retention of polar peptides.
  • Performs exceptionally well with LC-MS preferred mobile phases.
  • Offers high peak capacity for peptide mapping and complex sample separation.
  • The 4.6 x 50 mm column can separate 164 peaks with a resolution (Rs) of 1.
  • Suitable for routine and fast separations of 25–35 peptides.