Learn More
Waters Corp Waters BioSuite C18 PA-A Column, 100Å, 3 μm
Catalog No. 50781107 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioSuite Peptide Analysis columns are specifically optimized for peptide mapping from simple to complicated digests and primarily support legacy methods. Available in various configurations for LC or LC-MS applications, they offer excellent batch-to-batch reproducibility. This C18 3 μm PA-A column is a 100Å, difunctional bonded, low ligand density, silica-based reversed-phase column.
  • Primarily supports legacy methods.
  • Optimized for peptide mapping from simple to complicated digests.
  • Available in various configurations for LC or LC-MS applications, including 250 mm lengths for ultra high resolution.
  • Provides excellent batch-to-batch reproducibility for consistent results.
  • Uniquely QC tested specifically for peptide mapping.
  • A 100Å, difunctional bonded, low ligand density, silica-based reversed-phase column.
Catalog No. 50-781-107 Supplier Waters Corp Supplier No. 186002431
Add to Cart
Add to Cart
BioSuite Peptide Analysis columns are specifically optimized for peptide mapping from simple to complicated digests and primarily support legacy methods. Available in various configurations for LC or LC-MS applications, they offer excellent batch-to-batch reproducibility. This C18 3 μm PA-A column is a 100Å, difunctional bonded, low ligand density, silica-based reversed-phase column.
  • Primarily supports legacy methods.
  • Optimized for peptide mapping from simple to complicated digests.
  • Available in various configurations for LC or LC-MS applications, including 250 mm lengths for ultra high resolution.
  • Provides excellent batch-to-batch reproducibility for consistent results.
  • Uniquely QC tested specifically for peptide mapping.
  • A 100Å, difunctional bonded, low ligand density, silica-based reversed-phase column.