Learn More
Waters Corp Waters BioSuite CM Cation-exchange Column, 1000Å, 10 μm, 7.5 mm x 75 mm
Catalog No. 50780996 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioSuite CM Cation-Exchange (CXC) Column features a strong sulfopropyl ligand coupled to a pH-stable (pH 2 to 12) methacrylic ester-based polymeric resin. These ion-exchange columns are designed for protein and peptide separations, offering flexibility for chromatographers to develop methods based on minor charge differences.
  • Enables outstanding protein and peptide resolution and recovery.
  • Offers scalability from analytical to lab-scale preparative applications.
  • Incorporates high-performance chemistries for biomolecule isolation, analysis, and characterization.
  • Provides flexibility for separating proteins/peptides based on charge variations.
  • Available in nonporous and porous IEX options for varied resolution and binding capacity.
Catalog No. 50-780-996 Supplier Waters Corp Supplier No. 186002186
Add to Cart
Add to Cart
The BioSuite CM Cation-Exchange (CXC) Column features a strong sulfopropyl ligand coupled to a pH-stable (pH 2 to 12) methacrylic ester-based polymeric resin. These ion-exchange columns are designed for protein and peptide separations, offering flexibility for chromatographers to develop methods based on minor charge differences.
  • Enables outstanding protein and peptide resolution and recovery.
  • Offers scalability from analytical to lab-scale preparative applications.
  • Incorporates high-performance chemistries for biomolecule isolation, analysis, and characterization.
  • Provides flexibility for separating proteins/peptides based on charge variations.
  • Available in nonporous and porous IEX options for varied resolution and binding capacity.