Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters BioSuite C18 Column, 500Å, 13 μm, 21.5 mm × 150 mm

Catalog No. 50780965 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The BioSuite C18 Column supports legacy methods, offering a large pore size and high chemical stability. Its pC18 and pPhenyl resin-based packings are bonded to a pH stable, methacrylic ester-based polymeric resin. The 500 Å pore size accommodates proteins up to 2,500,000 Daltons. These columns provide superior chromatographic resolution, suitable for both “lab scale” isolations and narrow-bore HPLC/LC-MS applications.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Polymethacrylate
  • pH range: 2 pH to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Particle size: 13 μm
  • Pore size: 500 Å
  • Inner diameter: 21.5 mm
  • Length: 150 mm
  • Application: Peptide

Catalog No. 50-780-965 Supplier Waters Corp Supplier No. 186002155
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The BioSuite C18 Column supports legacy methods, offering a large pore size and high chemical stability. Its pC18 and pPhenyl resin-based packings are bonded to a pH stable, methacrylic ester-based polymeric resin. The 500 Å pore size accommodates proteins up to 2,500,000 Daltons. These columns provide superior chromatographic resolution, suitable for both “lab scale” isolations and narrow-bore HPLC/LC-MS applications.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Polymethacrylate
  • pH range: 2 pH to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Particle size: 13 μm
  • Pore size: 500 Å
  • Inner diameter: 21.5 mm
  • Length: 150 mm
  • Application: Peptide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.