Learn More
Waters Corp Waters BioSuite C18 Column, 500Å Pore, 7μm Particle, 4.6 mm × 150 mm
Catalog No. 50780964 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for the separation and analysis of biomolecules via high-performance liquid chromatography (HPLC).
  • Features C18 stationary phase for hydrophobic selectivity.
  • Has a 7 μm particle size and a 500Å pore size.
  • Dimensions: 4.6 mm diameter × 150 mm length.
  • Suitable for biomolecular weights from 1 KDa to 30 KDa.
  • Operates up to 6000 psi (414 bar) maximum pressure.
  • Ensures efficient separations, optimal sample penetration, and enhanced sensitivity.
  • Delivers excellent resolution, peak shapes, and reproducibility.
  • Features pH stable, methacrylic ester-based polymeric resin.
  • Primarily supports legacy methods.
Catalog No. 50-780-964 Supplier Waters Corp Supplier No. 186002154
This column is designed for the separation and analysis of biomolecules via high-performance liquid chromatography (HPLC).
  • Features C18 stationary phase for hydrophobic selectivity.
  • Has a 7 μm particle size and a 500Å pore size.
  • Dimensions: 4.6 mm diameter × 150 mm length.
  • Suitable for biomolecular weights from 1 KDa to 30 KDa.
  • Operates up to 6000 psi (414 bar) maximum pressure.
  • Ensures efficient separations, optimal sample penetration, and enhanced sensitivity.
  • Delivers excellent resolution, peak shapes, and reproducibility.
  • Features pH stable, methacrylic ester-based polymeric resin.
  • Primarily supports legacy methods.