Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters BioSuite C18 Column, 500Å, 7 μm, 2 mm × 150 mm

Catalog No. 50780963 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The BioSuite C18 Column primarily supports legacy methods, featuring large pore size and high chemical stability. Its 500 Å pore size pC18 base matrix accommodates proteins up to 2,500,000 Daltons, providing greater protein or peptide binding capacity than nonporous chemistries.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Polymethacrylate
  • PH range: 2 to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Molecular weight range: 1,000 to 30,000
  • Particle shape: Spherical
  • Particle size: 7 μm
  • Endfitting type: Parker-style
  • Pore size: 500 Å
  • Format: Column
  • System: HPLC
  • USP classification: L1
  • Inner diameter: 2 mm
  • Length: 150 mm
  • Application: Peptide
  • Units per package: 1 per package

Catalog No. 50-780-963 Supplier Waters Corp Supplier No. 186002153
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BioSuite C18 Column primarily supports legacy methods, featuring large pore size and high chemical stability. Its 500 Å pore size pC18 base matrix accommodates proteins up to 2,500,000 Daltons, providing greater protein or peptide binding capacity than nonporous chemistries.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Polymethacrylate
  • PH range: 2 to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Molecular weight range: 1,000 to 30,000
  • Particle shape: Spherical
  • Particle size: 7 μm
  • Endfitting type: Parker-style
  • Pore size: 500 Å
  • Format: Column
  • System: HPLC
  • USP classification: L1
  • Inner diameter: 2 mm
  • Length: 150 mm
  • Application: Peptide
  • Units per package: 1 per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.