Learn More
Waters Corp Waters Corporation BioSuite Phenyl Column, 10 μm, 1000 Å, 7.5 mm x 75 mm
Catalog No. 50780969 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Phenyl HIC columns provide an alternative to reversed-phase chromatography (RPC) for proteins and peptides sensitive to high organic solvents. Hydrophobic Interaction Chromatography (HIC) uses a weakly hydrophobic surface and salt gradient for precise, non-denaturing separation. The 10 μm column media features a phenyl group bonded to a methacrylic ester-based polymeric resin with a 1000 Å pore size, accommodating proteins up to 5,000,000 Daltons.
  • Separates proteins and peptides based on hydrophobic characteristics.
  • Offers a viable alternative to reversed-phase chromatography (RPC).
  • Combines non-denaturing characteristics with HPLC precision.
  • Phenyl group bonded to a methacrylic ester-based polymeric resin.
  • Large 1000 Å pore size.
  • Accommodates proteins up to 5,000,000 Daltons.
Catalog No. 50-780-969 Supplier Waters Corp Supplier No. 186002159
Add to Cart
Add to Cart
Phenyl HIC columns provide an alternative to reversed-phase chromatography (RPC) for proteins and peptides sensitive to high organic solvents. Hydrophobic Interaction Chromatography (HIC) uses a weakly hydrophobic surface and salt gradient for precise, non-denaturing separation. The 10 μm column media features a phenyl group bonded to a methacrylic ester-based polymeric resin with a 1000 Å pore size, accommodating proteins up to 5,000,000 Daltons.
  • Separates proteins and peptides based on hydrophobic characteristics.
  • Offers a viable alternative to reversed-phase chromatography (RPC).
  • Combines non-denaturing characteristics with HPLC precision.
  • Phenyl group bonded to a methacrylic ester-based polymeric resin.
  • Large 1000 Å pore size.
  • Accommodates proteins up to 5,000,000 Daltons.