Learn More
Waters Corp WATERS CORPORATION Column, Phenyl, 1000Å, 10 μm, 2 mm X 75 mm
Catalog No. 50780966 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed to support legacy methods and is suitable for applications where a large pore size and high chemical stability are important. It features a pH stable, methacrylic ester-based polymeric resin with a 1,000 Å pore size, capable of accommodating proteins up to 5,000,000 Daltons. The 2 mm x 75 mm format is ideal for narrow-bore HPLC and LC-MS applications.
  • Supports legacy methods.
  • Features large pore size and high chemical stability.
  • Utilizes a pH stable, methacrylic ester-based polymeric resin.
  • Accommodates proteins up to 5,000,000 Daltons.
  • Suitable for narrow-bore HPLC and/or LC-MS applications.
Catalog No. 50-780-966 Supplier Waters Corp Supplier No. 186002156
This column is designed to support legacy methods and is suitable for applications where a large pore size and high chemical stability are important. It features a pH stable, methacrylic ester-based polymeric resin with a 1,000 Å pore size, capable of accommodating proteins up to 5,000,000 Daltons. The 2 mm x 75 mm format is ideal for narrow-bore HPLC and LC-MS applications.
  • Supports legacy methods.
  • Features large pore size and high chemical stability.
  • Utilizes a pH stable, methacrylic ester-based polymeric resin.
  • Accommodates proteins up to 5,000,000 Daltons.
  • Suitable for narrow-bore HPLC and/or LC-MS applications.