Learn More
Waters Corp Waters BioSuite Phenyl Column, 1000 Å, 13 μm, 21.5 mm x 150 mm
Supplier: Waters Corp 186002158
This column supports legacy methods, and its large pore size and high chemical stability make it suitable for isolating and analyzing biocompounds. The pPhenyl RPC chemistry is available for “lab scale” isolations and narrow-bore HPLC or LC-MS applications.
- Chemistry: Phenyl
- Separation mode: Reversed phase
- Particle substrate: Polymethacrylate
- pH range: 2 to 12
- Maximum pressure: 6,000 psi (415 bar)
- Molecular weight range: 1,000 to 1,000,000 daltons
- Particle shape: Spherical
- Particle size: 13 μm
- Endfitting type: Parker-style
- Pore size: 1,000 Å (accommodates proteins up to 5,000,000 daltons)
- Format: Column
- System: HPLC
- USP classification: L11
- Inner diameter: 21.5 mm
- Length: 150 mm
- Application: Protein
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.