Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation BioSuite Q Anion-Exchange Column, 13 μm, 21.5 mm x 150 mm, Polymer

Catalog No. 50780988 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This BioSuite Q Anion-Exchange Column is designed for protein and peptide separations, offering excellent resolution and recovery. It is scalable from analytical to lab-scale preparative applications. With a 21.5 mm diameter, this column is suitable for preparative applications. The methacrylic ester-based polymeric resin provides pH stability from pH 2–12.

  • Designed for protein and peptide separations
  • Offers excellent resolution and recovery of proteins and peptides
  • Scalable from analytical to lab-scale preparative applications
  • Suitable for preparative applications due to 21.5 mm diameter
  • Methacrylic ester-based polymeric resin provides pH stability from pH 2–12
  • Strong-anion exchanger
  • Particle size: 13 μm
  • Dimensions: 21.5 mm x 150 mm

Catalog No. 50-780-988 Supplier Waters Corp Supplier No. 186002178
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This BioSuite Q Anion-Exchange Column is designed for protein and peptide separations, offering excellent resolution and recovery. It is scalable from analytical to lab-scale preparative applications. With a 21.5 mm diameter, this column is suitable for preparative applications. The methacrylic ester-based polymeric resin provides pH stability from pH 2–12.

  • Designed for protein and peptide separations
  • Offers excellent resolution and recovery of proteins and peptides
  • Scalable from analytical to lab-scale preparative applications
  • Suitable for preparative applications due to 21.5 mm diameter
  • Methacrylic ester-based polymeric resin provides pH stability from pH 2–12
  • Strong-anion exchanger
  • Particle size: 13 μm
  • Dimensions: 21.5 mm x 150 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.