Learn More
Waters Corp Waters Anion-Exchange Column, PEEK, 4000Å, 10μm, 4.6 mm x 50 mm
Catalog No. 50780986 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These ion-exchange columns are for high-speed protein and peptide separations. They feature strong and weak anion-exchangers (AXC) bonded to a pH stable (pH 2 to 12) methacrylic ester-based polymeric resin, allowing methods for separating proteins and/or peptides based on minor charge differences. Selected columns are available with PEEK hardware.
  • Strong/weak anion-exchangers (AXC) bonded to pH stable (pH 2 to 12) methacrylic ester-based polymeric resin
  • Separates proteins/peptides based on minor charge differences
  • Nonporous and porous IEX options for resolution or binding capacity
  • Available with PEEK hardware
  • Ideal for protein and peptide separations, biomolecule characterization
  • Suitable for oligonucleotide and gene therapeutics applications
Catalog No. 50-780-986 Supplier Waters Corp Supplier No. 186002176
Add to Cart
Add to Cart
These ion-exchange columns are for high-speed protein and peptide separations. They feature strong and weak anion-exchangers (AXC) bonded to a pH stable (pH 2 to 12) methacrylic ester-based polymeric resin, allowing methods for separating proteins and/or peptides based on minor charge differences. Selected columns are available with PEEK hardware.
  • Strong/weak anion-exchangers (AXC) bonded to pH stable (pH 2 to 12) methacrylic ester-based polymeric resin
  • Separates proteins/peptides based on minor charge differences
  • Nonporous and porous IEX options for resolution or binding capacity
  • Available with PEEK hardware
  • Ideal for protein and peptide separations, biomolecule characterization
  • Suitable for oligonucleotide and gene therapeutics applications