Learn More
Waters Corp Waters HPLC Guard Column, BioSuite SEC/GPC, 7.5 x 75 mm, 10 μm
Catalog No. 50780984 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioSuite Size-Exclusion (SEC) Columns are silica-based with diol chemistry, providing exceptional separation of molecules based on their size. They are designed for the confident isolation, analysis, and characterization of proteins and peptides, suitable for accurate molecular weight determinations. This guard column configuration is recommended to maximize the column life of analytical or preparative SEC columns.
  • Silica-based diol SEC columns provide exceptional separation of molecules based on size.
  • Optimizes column life of analytical or preparative SEC columns.
  • Utilizes rigid, yet wetable silica-based media.
  • Stable within a pH range of 2.5 to 7.5.
  • Designed for gel-permeation (GPC/SEC) mode applications.
  • Features a 7.5 × 75 mm dimension with 10 μm particle size.
Catalog No. 50-780-984 Supplier Waters Corp Supplier No. 186002174
BioSuite Size-Exclusion (SEC) Columns are silica-based with diol chemistry, providing exceptional separation of molecules based on their size. They are designed for the confident isolation, analysis, and characterization of proteins and peptides, suitable for accurate molecular weight determinations. This guard column configuration is recommended to maximize the column life of analytical or preparative SEC columns.
  • Silica-based diol SEC columns provide exceptional separation of molecules based on size.
  • Optimizes column life of analytical or preparative SEC columns.
  • Utilizes rigid, yet wetable silica-based media.
  • Stable within a pH range of 2.5 to 7.5.
  • Designed for gel-permeation (GPC/SEC) mode applications.
  • Features a 7.5 × 75 mm dimension with 10 μm particle size.