Learn More
Waters Corp Waters BioSuite SP Cation-exchange Column, 10 μm Particle, 1000 Å Pore, 7.5 × 75 mm
Supplier:  Waters Corp 186002184
Encompass_Preferred
This Cation-exchange Column is an HPLC column designed for assessing proteins and peptides based on their charge differences, utilizing separation chemistry specifically for cation-exchange chromatography. It features breakthrough separation technology for quicker, more precise conclusions about targeted peptides and proteins. The column is bonded to a pH stable (pH 2–12), methacrylic ester-based polymeric resin, allowing for flexibility in separating biomolecules with minor charge differences.
  • Features a cation exchanger as part of the separation chemistry.
  • Exchanger bonded to a pH stable methacrylic ester-based polymeric resin.
  • Enables quick and precise separations of proteins and peptides.
  • Offers flexibility to separate proteins and peptides based on very minor charge differences.
  • Usable pH range of 2–12.
Catalog No. 50-780-994