Learn More
Waters Corp Waters Cation-Exchange Column, 1300Å, 7 μm, 4.6 mm x 50 mm
Catalog No. 50780992 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters BioSuite™ Cation-Exchange Columns offer robust separation of proteins and peptides based on charge differences. These columns feature strong and weak cation-exchangers bonded to a pH stable (pH 2 to 12) methacrylic ester-based polymeric resin. Available in both nonporous and porous formats, they provide flexibility for method development in various applications.
  • pH range of 2 to 12.
  • Spherical particle shape.
  • Parker-style endfitting.
  • Compatible with HPLC systems.
  • Available with PEEK hardware.
  • Available in 21.5 mm preparative column sizes.
  • Designed for protein separation.
Catalog No. 50-780-992 Supplier Waters Corp Supplier No. 186002182
Waters BioSuite™ Cation-Exchange Columns offer robust separation of proteins and peptides based on charge differences. These columns feature strong and weak cation-exchangers bonded to a pH stable (pH 2 to 12) methacrylic ester-based polymeric resin. Available in both nonporous and porous formats, they provide flexibility for method development in various applications.
  • pH range of 2 to 12.
  • Spherical particle shape.
  • Parker-style endfitting.
  • Compatible with HPLC systems.
  • Available with PEEK hardware.
  • Available in 21.5 mm preparative column sizes.
  • Designed for protein separation.