Learn More
Carbosynth LLC. Biosynth 4-Methylumbelliferyl 2-Deoxy-2-Sulfamino-α-D-Glucopyranoside Sodium Salt, Fluorogenic Substrate, Heparan Sulphamidase, 500 ug
SDP
Catalog No. NC3929629
Encompass_Preferred
4-Methylumbelliferyl 2-deoxy-2-sulfamino-a-D-glucopyranoside sodium salt is a fluorogenic substrate of heparan sulphamidase. It is utilized in biochemical research, particularly for diagnosing Mucopolysaccharidosis IIIA and other lysosomal disorders by detecting associated enzymatic deficiencies.
  • Fluorogenic substrate for heparan sulphamidase
  • Enables detection of enzymatic activity via emission maxima of 445-454 nm
  • High biochemical purity
  • Molecular weight of 439.37 g/mol
  • Suitable for research use only
Catalog No. NC3929629 Supplier Carbosynth LLC. Supplier No. EM0660250MG
4-Methylumbelliferyl 2-deoxy-2-sulfamino-a-D-glucopyranoside sodium salt is a fluorogenic substrate of heparan sulphamidase. It is utilized in biochemical research, particularly for diagnosing Mucopolysaccharidosis IIIA and other lysosomal disorders by detecting associated enzymatic deficiencies.
  • Fluorogenic substrate for heparan sulphamidase
  • Enables detection of enzymatic activity via emission maxima of 445-454 nm
  • High biochemical purity
  • Molecular weight of 439.37 g/mol
  • Suitable for research use only