Learn More
Biosys Laboratories Biosys UriScan™ Urine Test Strips
SDP
Catalog No. 23679103
Change view
Click to view available options
Type:
10SGL Urine Test Strips
2GP Urine Test Strips
6L Urine Test Strips
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-679-103 6L Urine Test Strips
23-679-104 2GP Urine Test Strips
23-679-100 10SGL Urine Test Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-679-103 Supplier Biosys Laboratories Supplier No. U25
Add to Cart
Add to Cart
For measuring different parameters in urine sample
Specifications
Detectable Analytes Blood, Protein, Nitrite, pH, Glucose, Leukocyte
Type 6L Urine Test Strips