Learn More
Bioland Scientific LLC BioT (10 ml) up to 800 transfections
SDP
Catalog No. NC3740639
Encompass
This bulk volume of BioT provides up to 800 transfections in 10-cm dish or 330 transfections in T150 flask.
Catalog No. NC3740639 Supplier Bioland Scientific LLC Supplier No. B0103
This bulk volume of BioT provides up to 800 transfections in 10-cm dish or 330 transfections in T150 flask.