Learn More
Sigma Aldrich Fine Chemicals Biosciences BiotainerR media bottles c
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z3770821CS
Encompass
featuring a square footprint for space saving a wide neck for smoother sample pouring and a drip control ring. Also each bottle has easy-to-read graduations and an indented ribbed hand-grip for easier handling. The lot number and sterility dating are printed on each bottle for traceability.PETG bottles are suited for temperatures of -40 C to 60 C and the screw caps are fitted with high density polyethylene cap liners. Polycarbonate bottles can be heated to autoclaving temperatures and frozen to -100 C. The screw caps are fitted with silicone rubber laminated liners. The HDPE bottle is excellent for low-temperature freezing of media and similar products.
Catalog No. 50-290-5894