Learn More
Cole-Parmer EMD Millipore HYCON 941605 Blank Agar Strips; 50/Box
Supplier:  Cole-Parmer 8650960
Encompass
Compatible with the Biotest Hycon RCS Air Sampler 86509-00, the sampler operates on impaction principle, drawing airborne microorganisms onto an agar media strip for later incubation and analysis.
Catalog No. NC0228642