Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Biotic Index Data Sheet for Leaf Pack, Laminated
SDPSCI_ED

Catalog No. S81343 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
6/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81343 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81343 Supplier LaMotte™ Supplier No. 5882DATA6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water resistant data sheets for recording data after sorting and identifying freshwater aquatic macroinvertebrates.

  • Identification to Order and some Families
  • Includes instuctions and formulas for calculating the Pollution Tolerance Index (PTI) Score and PTI Rating
  • Write on, wipe off and reuse
  • English and Spanish

Specifications

Grade 3+
Quantity 6/Pk.
Product Type Laminated Data Sheet, Reusable
For Use With (Application) Identification of Aquatic Macroinvertebrates
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.