Learn More
Apexbio Technology LLC Biotin, 10g. Cas: 58-85-5 MFCD: MFCD00005541. Lysine probe coupled to mass spectrometry detection
SDP
Supplier:  Apexbio Technology LLC A801010000
Encompass_Preferred
NHS-biotin modification as a specific lysine probe coupled to mass spectrometry detection is increasingly used over the past years for assessing amino acid accessibility of proteins or complexes as an alternative when well-established methods are challenged1. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4981