Learn More
Apexbio Technology LLC Biotin-11-CTP 1ml (100 mM)
SDP
Catalog No. 503107125
Encompass_Preferred
Biotin-11-CTP is a substrate for SP6 T3 and T7 RNA polymerases It can substitute CTP in RNA labeling reactions with biotin during in vitro transcription Biotin-labeled RNA probes can be detected using streptavidin conjugates and are widely utilized in many molecular biology applications such as in situ hybridization and gene expression microarrays
Catalog No. 50-310-7125 Supplier Apexbio Technology LLC Supplier No. B88021
Biotin-11-CTP is a substrate for SP6 T3 and T7 RNA polymerases It can substitute CTP in RNA labeling reactions with biotin during in vitro transcription Biotin-labeled RNA probes can be detected using streptavidin conjugates and are widely utilized in many molecular biology applications such as in situ hybridization and gene expression microarrays