Learn More
Aat Bioquest Biotin-11-dATP (25 nmoles)
SDP
Catalog No. 502249670
Encompass_Preferred
The biotin-modified deoxyadenosine 5-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation random prime labeling cDNA labeling and 3-end labeling. This catalog is for 25 nmoles.
This item is not returnable. View return policy
Catalog No. 50-224-9670 Supplier Aat Bioquest Supplier No. 17014
The biotin-modified deoxyadenosine 5-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation random prime labeling cDNA labeling and 3-end labeling. This catalog is for 25 nmoles.