Learn More
Biotium Biotin-11-dCTP, 1mM in pH 7.5 TE buffer
Catalog No. 501964639
Encompass_Preferred
Biotin-11-dCTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3’-end terminal labeling. The number “11” is the number of atoms in the linker between biotin and dCTP. Properties: Colorless liquid; Store dessicated at -20⁰C; C28H42N7O16P3SLi4; MW: 885.4.
Catalog No. 50-196-4639 Supplier Biotium Supplier No. 40036
Biotin-11-dCTP can be enzymatically incorporated into DNA via nick translation, random priming, or 3’-end terminal labeling. The number “11” is the number of atoms in the linker between biotin and dCTP. Properties: Colorless liquid; Store dessicated at -20⁰C; C28H42N7O16P3SLi4; MW: 885.4.