Learn More
Aat Bioquest Biotin-11-dGTP 1 mM in Tris Buffer pH 7.5 (25 nmoles)
SDP
Catalog No. 502249671
Encompass_Preferred
The biotin-modified deoxyguanosine 5-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation random prime labeling cDNA labeling and 3-end labeling. This catalog is for 25 nmoles.
This item is not returnable. View return policy
Catalog No. 50-224-9671 Supplier Aat Bioquest Supplier No. 17015
The biotin-modified deoxyguanosine 5-triphosphates are widely used for a variety of non-radioactive DNA labeling reactions including nick translation random prime labeling cDNA labeling and 3-end labeling. This catalog is for 25 nmoles.