Learn More
Active Motif Biotin-14-dATP
SDP
Catalog No. NC1841519
Encompass
Active Motif's Active Motif's Biotin-14-dATP for biotin incorporation into DNA samples. It is used in nick translation to perform direct enzymatic labeling of DNA with biotin-14-ATP.
Catalog No. NC1841519 Supplier Active Motif Supplier No. 14138
Active Motif's Active Motif's Biotin-14-dATP for biotin incorporation into DNA samples. It is used in nick translation to perform direct enzymatic labeling of DNA with biotin-14-ATP.