Learn More
Axxora LLC Biotin-14-dATP, S pack, 200 uL
SDP
Supplier:  Axxora LLC JBSNU835BIO14S
Encompass
Application Notes: Incorporation into DNA/cDNA by - Nick Translation with DNAse I/ DNA, Polymerase I, Primer Extension with Klenow fragment.
Catalog No. NC1357968