Learn More
Aat Bioquest Biotin-14-dCTP 1 mM in Tris Buffer pH 7.5 (25 nmoles)
SDP
Catalog No. 502249675
Encompass_Preferred
The biotin-modified dCTP analogs are widely used for a variety of non-radioactive DNA labeling reactions including nick translation random prime labeling cDNA labeling and 3-end labeling. This catalog is for 25 nmoles.
This item is not returnable. View return policy
Catalog No. 50-224-9675 Supplier Aat Bioquest Supplier No. 17019
The biotin-modified dCTP analogs are widely used for a variety of non-radioactive DNA labeling reactions including nick translation random prime labeling cDNA labeling and 3-end labeling. This catalog is for 25 nmoles.