Learn More
Apexbio Technology LLC Biotin-16-dCTP,10ul (5 mM)
SDP
Catalog No. 502240007
Encompass_Preferred
Biotin-16-dCTP is a modified deoxynucleoside triphosphate (dNTP), serving as a convenient tool for the enzymatic introduction of the biotin moiety into a nucleic acid target of interest. The biotin moiety exhibits a very strong affinity for streptavidin. The strength and specificity of this interaction has been exploited in a variety of biological applications, such as secondary label introduction and affinity isolation. For the biotinylated nucleotides, biotin-16-dCTP and biotin-16-dUTP, strong amplicon formation can be observed with modified nucleotides substituted for 75% of their natural counterparts (i.e. dCTP and dUTP). Compared with biotin-16-dUTP whose yield decreases quickly above 75% substitution, the yield of biotin-16-dCTP decreases at a much higher substitution of ∼90%. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0007 Supplier Apexbio Technology LLC Supplier No. B81510.01
Biotin-16-dCTP is a modified deoxynucleoside triphosphate (dNTP), serving as a convenient tool for the enzymatic introduction of the biotin moiety into a nucleic acid target of interest. The biotin moiety exhibits a very strong affinity for streptavidin. The strength and specificity of this interaction has been exploited in a variety of biological applications, such as secondary label introduction and affinity isolation. For the biotinylated nucleotides, biotin-16-dCTP and biotin-16-dUTP, strong amplicon formation can be observed with modified nucleotides substituted for 75% of their natural counterparts (i.e. dCTP and dUTP). Compared with biotin-16-dUTP whose yield decreases quickly above 75% substitution, the yield of biotin-16-dCTP decreases at a much higher substitution of ∼90%. Other sizes are also available. Please inqury us for quote.