Learn More
Apexbio Technology LLC Biotin-16-dCTP 200ul (5 mM)
SDP
Catalog No. 5000063676
Encompass_Preferred
Biotin-16-dCTP is a biotinylated derivative of deoxycytidine triphosphate (dCTP) designed for enzymatic incorporation into nucleic acids during polymerase-mediated processes Structurally it consists of a dCTP core conjugated via a 16-atom spacer arm to a biotin moiety facilitating effective interaction with streptavidin This specific biotin-streptavidin binding affinity enables applications in nucleic acid labeling detection and affinity purification protocols PCR experiments indicate that biotin-16-dCTP incorporation by Taq polymerase can reach approximately 90% substitution without considerable reduction in amplification yield demonstrating greater polymerase compatibility compared to biotin-16-dUTP analogues
Catalog No. 50-000-63676 Supplier Apexbio Technology LLC Supplier No. B81510.2
Biotin-16-dCTP is a biotinylated derivative of deoxycytidine triphosphate (dCTP) designed for enzymatic incorporation into nucleic acids during polymerase-mediated processes Structurally it consists of a dCTP core conjugated via a 16-atom spacer arm to a biotin moiety facilitating effective interaction with streptavidin This specific biotin-streptavidin binding affinity enables applications in nucleic acid labeling detection and affinity purification protocols PCR experiments indicate that biotin-16-dCTP incorporation by Taq polymerase can reach approximately 90% substitution without considerable reduction in amplification yield demonstrating greater polymerase compatibility compared to biotin-16-dUTP analogues