Learn More
Biotium Biotin-16-UTP, 10 mM in pH 7.5 Tris-HCl Solution
Catalog No. 501964632
Encompass_Preferred
Biotin-16-UTP can be enzymatically incorporated into RNA. The number “16” is the number of atoms in the linker between biotin and UTP. The length of the linker affects the incorporation efficiency of the biotin-UTP probe into RNA using RNA polymerases, and it also affects biotin/avidin or biotin/streptavidin. In general, the shorter the linker, the more efficiently the biotin-UTP is incorporated into RNA by RNA polymerases. On the other hand, the longer the linker, the better biotin can interact with avidin or streptavidin. Properties: Colorless liquid; Store at -20⁰C; C32H48N7O19P3SLi4; MW: 987.51.
Catalog No. 50-196-4632 Supplier Biotium Supplier No. 40023
Biotin-16-UTP can be enzymatically incorporated into RNA. The number “16” is the number of atoms in the linker between biotin and UTP. The length of the linker affects the incorporation efficiency of the biotin-UTP probe into RNA using RNA polymerases, and it also affects biotin/avidin or biotin/streptavidin. In general, the shorter the linker, the more efficiently the biotin-UTP is incorporated into RNA by RNA polymerases. On the other hand, the longer the linker, the better biotin can interact with avidin or streptavidin. Properties: Colorless liquid; Store at -20⁰C; C32H48N7O19P3SLi4; MW: 987.51.