Learn More
Apexbio Technology LLC Biotin-16-UTP 200ul (10 mM)
SDP
Catalog No. 503107375
Encompass_Preferred
Biotin-16-UTP is a biotin-labeled uridine triphosphate used for RNA detection and purification The biotin label allows RNA molecules to bind with streptavidin or anti-biotin proteins facilitating specific RNA detection and purification In in vitro transcription reactions Biotin-16-UTP can be incorporated into RNA molecules generating biotin-labeled RNA This labeled RNA has wide applications in RNA-protein interaction studies RNA detection and RNA purification Additionally biotin-labeled RNA can be applied in in vivo experiments to investigate RNA localization and function within cells
Catalog No. 50-310-7375 Supplier Apexbio Technology LLC Supplier No. B81540.2
Biotin-16-UTP is a biotin-labeled uridine triphosphate used for RNA detection and purification The biotin label allows RNA molecules to bind with streptavidin or anti-biotin proteins facilitating specific RNA detection and purification In in vitro transcription reactions Biotin-16-UTP can be incorporated into RNA molecules generating biotin-labeled RNA This labeled RNA has wide applications in RNA-protein interaction studies RNA detection and RNA purification Additionally biotin-labeled RNA can be applied in in vivo experiments to investigate RNA localization and function within cells