Learn More
U.S. Pharmacopeia Biotin, 58-85-5, MFCD00005541, 200 mg
Catalog No. NC0262716
Encompass
Empirical Formula (Hill Notation): C10H16N2O3S, Molecular Weight: 244.31, MP: 231-233 °C, Synonym: D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H.
Catalog No. NC0262716 Supplier U.S. Pharmacopeia Supplier No. 1071508
Empirical Formula (Hill Notation): C10H16N2O3S, Molecular Weight: 244.31, MP: 231-233 °C, Synonym: D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H.