Learn More
Sigma Aldrich Fine Chemicals Biosciences Biotin 2233 d4 98 atom D1MG
Catalog No. 502905116
Encompass
Biotin is a water-soluble vitamin B which is also known as vitamin B7. It is involved in the synthesis of fatty acids and gluconeogenesis. Biotin is found in plants and animals. Its deficiency causes retardation in growth alopecia and neurological impairment in humans and animals.Biotin-2AE 2AE 3AE 3AE-d4 is an isotope of vitamin B7 wherein C-2AE C-2AE C-3AE C-3AE protons are replaced by deuterium.
Catalog No. 50-290-5116 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8096081MG
Biotin is a water-soluble vitamin B which is also known as vitamin B7. It is involved in the synthesis of fatty acids and gluconeogenesis. Biotin is found in plants and animals. Its deficiency causes retardation in growth alopecia and neurological impairment in humans and animals.Biotin-2AE 2AE 3AE 3AE-d4 is an isotope of vitamin B7 wherein C-2AE C-2AE C-3AE C-3AE protons are replaced by deuterium.