Learn More
Sigma Aldrich Fine Chemicals Biosciences Biotin 3-sulfo-N-hydroxysu10MG
Catalog No. 502898332
Encompass
Biotinavidin second antibody signal systems involve the conjugation of biotin to a secondary antibody. Once the secondary antibody binds to a target primary antibody in an immunochemical assay the signal is developed by allowing avidin which has been conjugated to an enzyme or signal molecule to bind to the antibody associated biotin molecule.
Catalog No. 50-289-8332 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B516110MG
Biotinavidin second antibody signal systems involve the conjugation of biotin to a secondary antibody. Once the secondary antibody binds to a target primary antibody in an immunochemical assay the signal is developed by allowing avidin which has been conjugated to an enzyme or signal molecule to bind to the antibody associated biotin molecule.