Learn More
TARGETMOL CHEMICALS INC Biotin-4-aminophenol 10MG
SDP
Catalog No. 502879841
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Biotin-4-aminophenol (BP) is a selective and potent biotin-phenol analog that works by generating free radicals and binding to tyrosine residues in proteins. Purity 98.99%
Catalog No. 50-287-9841 Supplier TARGETMOL CHEMICALS INC Supplier No. T6478110MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Biotin-4-aminophenol (BP) is a selective and potent biotin-phenol analog that works by generating free radicals and binding to tyrosine residues in proteins. Purity 98.99%