Learn More
Apexbio Technology LLC BIOTIN-4-AMINOPHENOL-25MG
SDP
Catalog No. 5000569900
Encompass_Preferred
Biotin-4-aminophenol (CAS No 901770-40-9) is a chemical compound used in biomedical research originating from the field of biochemistry It operates primarily through biotinylation affecting pathways involved in cellular signaling and transport In vitro it demonstrates activity in the nanomolar to low micromolar range making it suitable for precise biological studies Typical applications include experiments in various cell models animal studies and drug screening processes where it serves as a critical reagent for labeling and tracking molecules Experimental concentrations typically depend on the specific research design allowing researchers to tailor its use to their particular study needs
Catalog No. 50-005-69900 Supplier Apexbio Technology LLC Supplier No. C821025
Biotin-4-aminophenol (CAS No 901770-40-9) is a chemical compound used in biomedical research originating from the field of biochemistry It operates primarily through biotinylation affecting pathways involved in cellular signaling and transport In vitro it demonstrates activity in the nanomolar to low micromolar range making it suitable for precise biological studies Typical applications include experiments in various cell models animal studies and drug screening processes where it serves as a critical reagent for labeling and tracking molecules Experimental concentrations typically depend on the specific research design allowing researchers to tailor its use to their particular study needs