Learn More
Medchemexpress LLC Biotin-4-aminophenol | 901770-40-9 | >99.45% | C16H21N3O3S | 1 ML
SDP
Catalog No. 5000353605
Encompass_Preferred
Biotin-4-aminophenol is a biotin-phenol analog that generates free radicals and conjugates to tyrosine residues in proteins. It is noted for its more efficient and selective conjugation compared to previously reported BP1. This product is intended for research use only.
  • Generates free radicals
  • Conjugates to tyrosine residues in proteins
  • More efficient and selective conjugation than BP1
  • For research use only
Catalog No. 50-003-53605 Supplier Medchemexpress LLC Supplier No. HY14189810MM/1ML
Biotin-4-aminophenol is a biotin-phenol analog that generates free radicals and conjugates to tyrosine residues in proteins. It is noted for its more efficient and selective conjugation compared to previously reported BP1. This product is intended for research use only.
  • Generates free radicals
  • Conjugates to tyrosine residues in proteins
  • More efficient and selective conjugation than BP1
  • For research use only