Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Biotin-4-aminophenol | 901770-40-9 | 99.45% | C16H21N3O3S | 100 MG
SDP

Catalog No. 5000354364
Encompass_Preferred

Biotin-4-aminophenol is a biotin-phenol analog that generates free radicals. It conjugates to tyrosine residues in proteins more efficiently and selectively than the previously reported BP1.

  • Biotin-phenol analog
  • Generates free radicals
  • Efficiently conjugates to tyrosine residues in proteins
  • More selective than BP1
  • Molecular weight: 335.42
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-54364 Supplier Medchemexpress LLC Supplier No. HY141898100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotin-4-aminophenol is a biotin-phenol analog that generates free radicals. It conjugates to tyrosine residues in proteins more efficiently and selectively than the previously reported BP1.

  • Biotin-phenol analog
  • Generates free radicals
  • Efficiently conjugates to tyrosine residues in proteins
  • More selective than BP1
  • Molecular weight: 335.42
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.