Learn More
Medchemexpress LLC Biotin-4-aminophenol | 901770-40-9 | 99.45% | C16H21N3O3S | 25 MG
SDP
Catalog No. 5000357223
Encompass_Preferred
Biotin-4-aminophenol is a biotin-phenol analog. It is designed for research use only and generates free radicals that conjugate to tyrosine residues in proteins more efficiently and selectively than previously reported BP1. This product is suitable for various laboratory applications.
  • Generates free radicals
  • Efficiently conjugates to tyrosine residues in proteins
  • More selective than BP1
  • For research use only
  • White to off-white solid appearance
Catalog No. 50-003-57223 Supplier Medchemexpress LLC Supplier No. 25MGHY141898
Biotin-4-aminophenol is a biotin-phenol analog. It is designed for research use only and generates free radicals that conjugate to tyrosine residues in proteins more efficiently and selectively than previously reported BP1. This product is suitable for various laboratory applications.
  • Generates free radicals
  • Efficiently conjugates to tyrosine residues in proteins
  • More selective than BP1
  • For research use only
  • White to off-white solid appearance